Anti-Cytokeratin 13 Mouse Monoclonal Antibody [clone: KS-1A3]
Supplier: Diagnostic Biosystems
MOB443-01-RUO MOB443-05-RUO MOB443-RUO
CA76633-928EA 144.79 CAD
CA76633-928 CA76633-930 CA76633-932
Anti-Cytokeratin 13 Mouse Monoclonal Antibody [clone: KS-1A3]
Antibodies
This antibody is specific to 54 kDa protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in squamous, non-keratinized epithelium, transitional epithelium and myoepithelium. This antibody is useful for the identification of carcinomas of trachea, sweat glands, bladder, ectocervix, tongue, esophagus, anal canal and the basal layer of keratinized epidermis.