Anti-MUM1 Protein Mouse Monoclonal Antibody [clone: MUM1p]
Supplier: Diagnostic Biosystems
MOB420-01-RUO MOB420-05-RUO MOB420-RUO
CA76633-844EA 299.77 CAD
CA76633-844 CA76633-846 CA76633-848
Anti-MUM1 Protein Mouse Monoclonal Antibody [clone: MUM1p]
Antibodies
Multiple myeloma oncogene-1 (MUM1) is a 50kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies.