Anti-Factor XIIIa Mouse Monoclonal Antibody [clone: AC-1A1]
Supplier: Diagnostic Biosystems
MOB321-01-RUO MOB321-05-RUO MOB321-RUO
CA76633-662EA 170.34 CAD
CA76633-662 CA76633-664 CA76633-666
Anti-Factor XIIIa Mouse Monoclonal Antibody [clone: AC-1A1]
Antibodies
This antibody is specific to 160 kDa protein known as Factor XIII-A. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and v variable reaction with these types of tumors.