Anti-CD79A B cell Mouse Monoclonal Antibody [clone: HM47/A9]
Supplier: Diagnostic Biosystems
MOB242-01-RUO MOB242-05-RUO MOB242-RUO
CA76633-504EA 206.11 CAD
CA76633-504 CA76633-506 CA76633-508
Anti-CD79A B cell Mouse Monoclonal Antibody [clone: HM47/A9]
Antibodies
This antibody is specific to human CD79a antigen complex, consisting of two polypeptides non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of polypeptide and immunoglobulins constitute the B cell antigen receptor. The two components of this complex are designated CD79a and CD79b. The CD79a antigen appears at the Pre-B cell stage, early in maturation and persists until the plasma cell stage. The CD79a antigen is found in majority of acute leukemias of precursor B cell type, in B cell lines and B cell lymphomas.