Anti-c-Myc Mouse Monoclonal Antibody [clone: 9 E10]
Supplier: Diagnostic Biosystems
MOB231-01-RUO MOB231-05-RUO MOB231-RUO
CA76633-486EA 144.79 CAD
CA76633-486 CA76633-488 CA76633-490
Anti-c-Myc Mouse Monoclonal Antibody [clone: 9 E10]
Antibodies
This antibody recognizes an epitope located in the amino acid residue 410 to 419 of human oncogene product c-Myc. This antibody reacts with both components of the P62 and P64 c-Myc.