Anti-Neurofilaments Mouse Monoclonal Antibody [clone: 2F11]
Supplier: Diagnostic Biosystems
MOB080-01-RUO MOB080-05-RUO MOB080-RUO
CA76633-238EA 144.79 CAD
CA76633-238 CA76633-240 CA76633-242
Anti-Neurofilaments Mouse Monoclonal Antibody [clone: 2F11]
Antibodies
This antibody reacts with 70 and 200 kDa component of the neurofilament polypeptide subunit. It stains neurons, neuronal processes and peripheral nerves. This antibody reacts with some neuroblastomas and gangliomas but not with other non-neuronal tumors.