Anti-Melanoma Mouse Monoclonal Antibody [clone: HMB45]
Supplier: Diagnostic Biosystems
MOB079-RUO MOB079-01-RUO MOB079-05-RUO
CA76633-236EA 912.96 CAD
CA76633-236 CA76633-232 CA76633-234
Anti-Melanoma Mouse Monoclonal Antibody [clone: HMB45]
Antibodies
This antibody reacts with a neuraminidase sensitive oligosaccharide side chain of a glycoconjugate present in immature melanosomes. It reacts with junctional and blue nevus cells. Non-melanocytic cells are negative.