Anti-CD21 Mouse Monoclonal Antibody [clone: 1F8]
Supplier: Diagnostic Biosystems
MOB028-01-RUO MOB028-05-RUO MOB028-RUO
CA76633-110EA 211.23 CAD
CA76633-110 CA76633-112 CA76633-114
Anti-CD21 Mouse Monoclonal Antibody [clone: 1F8]
Antibodies
This antibody reacts with CD21 antigen of MW 145 kDa, present on the surface of mature B cells. The antigen is present at high density on follicular dendritic cells. CD21 is involved in transmitting growth-promoting signals to the interior of the B cell.