Anti-CEA Mouse Monoclonal Antibody [clone: COL-1]
Supplier: Diagnostic Biosystems
MOB008-01-RUO MOB008-05-RUO MOB008-RUO
CA76633-104EA 144.79 CAD
CA76633-104 CA76633-106 CA76633-108
Anti-CEA Mouse Monoclonal Antibody [clone: COL-1]
Antibodies
This antibody is a member of a family of complex antigens, which also includes non-specific cross-reacting antigens (NCA). This antibody stains specific regions of CEA. It reacts with colorectal adenocarcinomas but does not show any staining with polymorphonuclear neutrophils and erythrocytes.