Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Supplier: Diagnostic Biosystems
MOB111-01-RUO MOB111-05-RUO MOB111-RUO
CA76633-310EA 144.79 CAD
CA76633-310 CA76633-312 CA76633-314
Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Antibodies
This antibody is specific against an epitope located on the β chain (i.e., in S-100a and S-100b) but not on the α chain of S-100 (i.e., in S-100a and S-100ao). This antibody can be used to localize S-100a and S-100b in various tissue sections.