Anti-CMV Cocktail Mouse Monoclonal Antibody [clone: DDG9 + CCH1]
Supplier: Diagnostic Biosystems
MOB049-01-RUO MOB049-05-RUO MOB049-RUO
CA76633-152EA 211.23 CAD
CA76633-152 CA76633-154 CA76633-156
Anti-CMV Cocktail Mouse Monoclonal Antibody [clone: DDG9 + CCH1]
Antibodies
This cocktail antibody stains infected cells giving a nuclear staining pattern with an immediate early and an early CMV antigen.