Anti-CD45 LCA Mouse Monoclonal Antibody [clone: 2B11+PD7/26]
Supplier: Diagnostic Biosystems
MOB040-05-RUO MOB040-RUO MOB040-01-RUO
CA76633-136EA 316.81 CAD
CA76633-136 CA76633-138 CA76633-134
Anti-CD45 LCA Mouse Monoclonal Antibody [clone: 2B11+PD7/26]
Antibodies
CD45 is useful in differential diagnosis of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.