Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Supplier: Diagnostic Biosystems
MOB349-05-RUO MOB349-RUO MOB349-01-RUO
CA76633-736EA 926.58 CAD
CA76633-736 CA76633-738 CA76633-734
Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Antibodies
This antibody reacts with an unknown antigen on microvilli of mesothelioma cells. It stains normal mesothelial cells as well as epithelial mesotheliomas in a thick membrane pattern due to abundant lung microvilli on the surface of these cells.