Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: KP1]
Supplier: Diagnostic Biosystems
MOB167-01-RUO MOB167-05-RUO MOB167-RUO
CA76633-376EA 144.79 CAD
CA76633-376 CA76633-378 CA76633-380
Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: KP1]
Antibodies
This antibody is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as plasmacytoid T cells.