Anti-CD61 Platelet Glycoprotein IIIa Mouse Monoclonal Antibody [clone: Y2/51]
Supplier: Diagnostic Biosystems
MOB164-01-RUO MOB164-05-RUO MOB164-RUO
CA76633-370EA 144.79 CAD
CA76633-370 CA76633-372 CA76633-374
Anti-CD61 Platelet Glycoprotein IIIa Mouse Monoclonal Antibody [clone: Y2/51]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa molecular weight present on human platelets and reacts with megakaryocytes and megakaryoblasts. The antigen is also present on endothelium and osteoclasts. The glycoprotein IIIa molecule produced by endothelium cells is identical in sequence to platelet glycoprotein IIIa.