Anti-CD43 T cell Mouse Monoclonal Antibody [clone: DF-T1]

Supplier: Diagnostic Biosystems
MOB039-01-RUO MOB039-05-RUO MOB039-RUO
CA76633-128EA 144.79 CAD
CA76633-128 CA76633-130 CA76633-132
Anti-CD43 T cell Mouse Monoclonal Antibody [clone: DF-T1]
Antibodies
This antibody reacts with an antigen expressed by normal and neoplastic T cells and myeloid lineage cells. It strongly stains T cells in the interfollicular areas of normal lymphoid tissues.
Order Now

SPECIFICATIONS

Antigen name CD43 T cell
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone DF-T1
Reactivity Human
Host Mouse
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR