Anti-MOC-31 Mouse Monoclonal Antibody [clone: MOC-31]
Supplier: Diagnostic Biosystems
MOB546-01-RUO MOB546-05-RUO MOB546-RUO
CA76634-048EA 158.43 CAD
CA76634-048 CA76634-050 CA76634-052
Anti-MOC-31 Mouse Monoclonal Antibody [clone: MOC-31]
Antibodies
MOC-31 antibody recognizes an epithelial-associated, glycoprotein located on the cell membrane surface and in the cytoplasm of virtually all epithelial cells. It is not present in most squamous epithelia, hepatocytes, renal proximal tubular cells, gastric parietal cells and myoepithelial cells. MOC-31 may be used in a panel of antibodies as a negative marker for mesothelioma, or lung adenocarcinoma. Studies have shown that MOC31 is useful in differentiating tumors of unknown origin in liver cancers and distinguishing Cholangiocarcinoma from Hepatocellular Carcinomas.