Anti-TdT Mouse Monoclonal Antibody [clone: SEN 28]
Supplier: Diagnostic Biosystems
MOB545-01-RUO MOB545-05-RUO MOB545-RUO
CA76634-042EA 207.8 CAD
CA76634-042 CA76634-044 CA76634-046
Anti-TdT Mouse Monoclonal Antibody [clone: SEN 28]
Antibodies
This antibody stains a 60 kDa peptide in human lymphoblasts. TdT is present in the cells during immunoglobulin gene rearrangement and plays an important role in lymphocyte maturation. TdT marks both T and B immature lymphocytes normally found in thymus and bone marrow.