Anti-Tyrosinase Mouse Monoclonal Antibody [clone: T311]
Supplier: Diagnostic Biosystems
MOB290-01-RUO MOB290-05-RUO MOB290-RUO
CA76633-602EA 178.86 CAD
CA76633-602 CA76633-604 CA76633-606
Anti-Tyrosinase Mouse Monoclonal Antibody [clone: T311]
Antibodies
This antibody is specific to a cluster of proteins between 70 to 80 kDa known as tyrosinase. This antibody does not show any cross-reaction with MAGE-1 and tyrosinase-related protein-1, TRP-1/gp75. Staining of melanomas with this antibody showed tyrosinase in melanotic as well as amelanotic variants.