Anti-CD99 Mouse Monoclonal Antibody [clone: HO36-1.1]
Supplier: Diagnostic Biosystems
MOB262-01-RUO MOB262-05-RUO MOB262-RUO
CA76633-540EA 151.62 CAD
CA76633-540 CA76633-542 CA76633-544
Anti-CD99 Mouse Monoclonal Antibody [clone: HO36-1.1]
Antibodies
This antibody is specific to a Sialoglycoprotein of 27 to 32 kDa known as CD99 or MIC2 gene product. CD99 is expressed on the cell membrane of some lymphocytes, cortical thymocytes and granulosa cells of the ovary, Sertoli cells of testis, pancreatic islet cells and some endothelial cells. Mature granulocytes express very little or no CD99.