Anti-CD44 HCAM Mouse Monoclonal Antibody [clone: 156-3C11]
Supplier: Diagnostic Biosystems
MOB256-01-RUO MOB256-05-RUO MOB256-RUO
CA76633-528EA 158.43 CAD
CA76633-528 CA76633-530 CA76633-532
Anti-CD44 HCAM Mouse Monoclonal Antibody [clone: 156-3C11]
Antibodies
This antibody is specific to a cell surface glycoprotein of 80 to 95 kDa, known as CD44. CD44 is expressed on the surface of lymphocytes, monocytes and granulocytes. This antibody reacts with red cells pretreated with 2-Aminoethyl Isothiouronium Bromide (AET).