Anti-CD57 HNK-1 Mouse Monoclonal Antibody [clone: NK-1]

Supplier: Diagnostic Biosystems
MOB163-01-RUO MOB163-05-RUO MOB163-RUO
CA76633-364EA 144.79 CAD
CA76633-364 CA76633-366 CA76633-368
Anti-CD57 HNK-1 Mouse Monoclonal Antibody [clone: NK-1]
Antibodies
This antibody is specific to a 110 kDa human CD57 myeloid associated glycoprotein. It stains mononuclear cells with natural killer activity, neuroendocrine cells and their tumors.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name CD57 hnk-1
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone NK-1
Reactivity Human
Host Mouse
Isotype IgM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR