Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Supplier: Diagnostic Biosystems
MOB129-01-RUO MOB129-05-RUO MOB129-RUO
CA76633-352EA 144.79 CAD
CA76633-352 CA76633-354 CA76633-356
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Antibodies
Alpha Fetoprotein (AFP) is a single chain glycoprotein of 70 kDa that is normally expressed in the fetal liver, gastrointestinal tract and yolk sac. This antibody recognizes an epitope of α-fetoprotein present in human and several other species.