Anti-Actin Sarcomeric Mouse Monoclonal Antibody [clone: 5C5]
Supplier: Diagnostic Biosystems
MOB128-01-RUO MOB128-05-RUO MOB128-RUO
CA76633-346EA 144.79 CAD
CA76633-346 CA76633-348 CA76633-350
Anti-Actin Sarcomeric Mouse Monoclonal Antibody [clone: 5C5]
Antibodies
This antibody is specific to α-smooth muscle isoform of actin. It reacts with smooth muscle cells of vessels and different parenchymes.