Anti-Cytokeratin AE3 Basic Mouse Monoclonal Antibody [clone: AE3]
Supplier: Diagnostic Biosystems
MOB093-01-RUO MOB093-05-RUO MOB093-RUO
CA76633-280EA 155.01 CAD
CA76633-280 CA76633-282 CA76633-284
Anti-Cytokeratin AE3 Basic Mouse Monoclonal Antibody [clone: AE3]
Antibodies
This antibody recognizes basic (Type II or HMW) cytokeratins, which includes 67 kDa (CK1), 64 kDa (CK3), 59 kDa (CK4), 58 kDa (CK5), 56 kDa (CK6), 52 kDa (CK8).