Anti-Cytokeratin 8 Mouse Monoclonal Antibody [clone: 35bH11]
Supplier: Diagnostic Biosystems
MOB054-01-RUO MOB054-05-RUO MOB054-RUO
CA76633-164EA 144.79 CAD
CA76633-164 CA76633-166 CA76633-168
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [clone: 35bH11]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains strongly with non-squamous epithelium but does not stain squamous epithelium. It reacts with the sweat gland and ducts in the skin. Adenocarcinomas of the ovary, gastrointestinal tract, ductal carcinoma of breast, pancreas and bile ducts, stain positive with this antibody.