Anti-Actin Muscle Mouse Monoclonal Antibody [clone: HHF35]
Supplier: Diagnostic Biosystems
MOB002-01-RUO MOB002-05-RUO MOB002-RUO
CA76633-080EA 144.79 CAD
CA76633-080 CA76633-082 CA76633-084
Anti-Actin Muscle Mouse Monoclonal Antibody [clone: HHF35]
Antibodies
This pan actin is useful in identifying tumors arising from muscle cells, i.e., leiomyosarcoma as well as rhabdomyosarcoma.