Slide Drying Hotplate, Mopec
Supplier: Mortech Manufacturing
BK400 BK400-L
76579-276EA 3831.42 CAD
76579-276 76579-278
Slide Drying Hotplate, Mopec
Slide Warmers/Dryers
Mopec’s slide drying hotplate is designed to quickly and efficiently dry paraffin tissue sections. The rapid heating system ensures the hotplate is ready to use in less time, while the digital temperature control allows for accurate control of temperatures.
  • Designed to take up minimal bench-top space
  • Digital temperature control to meet precise temperature requirements
  • Rapid heating system ensures system is ready to use in less time
  • Slide support rails can be adjusted to fit different slide sizes
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps reduce drying time and improve temperature control (BK400-L)
Slides are places on the four moveable rails at an angle to allow water drain off, while heated air passes underneath the slides to reduce drying times. The system features an optional lid to reduce drying times and improve temperature control. Made from stainless steel and can be used while slides are on the hotplate.