Heated Forceps System, Mopec
Supplier: Mortech Manufacturing
BK300-SE BK300-SM
76579-222EA 4131.27 CAD
76579-222 76579-224
Heated Forceps System, Mopec
Forceps
Mopec’s heated forceps system is designed to take up minimal bench-top space and comes complete with a power supply, control unit, and two different heated forceps. The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth.
  • Choose either serrated or smooth tips
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth
Sold individually.
Delivery information: System includes:
1 mm Heated forceps (Red)
2 mm Heated forceps (Yellow)
BK330: Mopec control unit
BK320: Mopec power supply