Formaway Green Liquid Formalin Neutralizer, Mopec
Supplier: MORTECH MANUFACTURING
BG560-1 BG560-2.5 BG560-5
76579-378EA 139.99 CAD
76579-378 76579-380 76579-382
Formaway Green Liquid Formalin Neutralizer, Mopec
Stains Microscopy and staining reagents
Mopec’s Formaway green liquid formalin neutralizer quickly and effectively neutralizes spent formalin for sewer disposal. Simply add 32 ounces of Formaway green to each gallon of 10% neutral buffered formalin waste and allow to stand for at least 4 hours before disposing.
  • Sold individually
Collect formalin waste in the Formaway collection container or equivalent container. Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste formalin. Add 32-fl oz. of Formaway green to each gallon (128-fl oz.) of 10% NBF formalin waste. (For metric users, add 1 liter of Formaway green to 4 liters of 10% NBF formalin waste.) Secure lid on waste collection container and agitate container to mix solutions. Allow mixed solutions to stand for approximately 4 hours, adequately reducing the formalin in the waste solution to a non hazardous safe disposal into the sanitary sewer. Repeat step #4, decreasing any potential exposure to the headspace vapor in the collection container. Test treated solution with rapid QC analysis and carefully pour treated waste into the sanitary sewer. Rinse container with cold tap water, preparing it for re-use.
Order Now
SPECIFICATIONS
Environmentally Preferable