Coldplate, Mopec
Supplier: Mortech Manufacturing
BK600 BK600-B BK600-C
76579-290EA 955.93 CAD
76579-290 76579-294 76579-292
Coldplate, Mopec
Coolers Benchtop Coolers
The Mopec Coldplate is designed to replace chilling sprays or ice blocks that are typically used to cool down tissue blocks before sectioning.
  • 36 tissue block capacity
  • Designed to take up minimal bench-top space
  • Removes the need for chilling sprays or ice blocks
  • Remains cold for up to 4 hours
  • No electrical components or compressors required
Featuring a large work surface, the insulated base and aluminum coldplate allow for up to 36 blocks to be cooled at one time. The base will collect any condensation created, keeping the bench and floor dry. Store in a freezer compartment when not in use.