Formalin prefilled 1:10 Bulk Containers

Supplier: MORTECH MANUFACTURING
Warning

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

BG520 BG525 BG530 BG555 BG501
76579-336EA 115.42 CAD
76579-336 76579-340 76579-342 76579-346 76579-314
Formalin prefilled 1:10 Bulk Containers
Formalin

Mopec’s Bulk Formalin Containers are filled with a 10% neutral buffered formalin used as a general purpose fixative for tissues. Penetrates quickly, but fixes slowly. No other dilution or addition of other agents is necessary. Comes in a variety of sizes.


  • Filled with a 10% neutral buffered formalin
  • General purpose fixative for tissues.
  • Penetrates quickly, but fixes slowly
  • No other dilution or additional agents are necessary

Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR