Harmony Tissue Marking Dye Kit, Set of 7, Mortech
Supplier: MORTECH MANUFACTURING
BG403-250 BG403-60 BG403-60S
76549-754EA 1422.13 CAD
76549-754 76549-756 76549-758
Harmony Tissue Marking Dye Kit, Set of 7, Mortech
Stains Histology Stains
Mopec’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in nine colors, with a standardized viscosity over the range. This ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color.
  • Emulsified pigments are blended perfectly with an aqueous base to provide a consistent and high-quality product every time
  • Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
  • Dries in 3 minutes, improving laboratory workflow at the grossing station
  • Nonflammable
  • Available in round neck, induction sealed bottles with a standard flip top lid
  • Option to add BG200: Harmony Tissue Marking Dye Stand for 2 oz bottles
Harmony has also been purposely engineered to reduce in viscosity during application, giving users an easy to apply dye without any dripping or running and ensures an even coverage of dye on the specimen.
High concentrations of pigment in the dyes increase their durability and allow diagnostic specimens to be more easily marked. Harmony coats specimens with minimal penetration of the tissue surface, whilst its strong adhesion qualities mean the marking dye will not interfere with diagnostic interpretations, making it ideal for radical prostatectomies.
Delivery information: Includes (1) bottle of each color: red, orange, yellow, green, blue, violet, and black.