Disposable Autopsy and Pathology Grossing Prep Knives , Mortech
Supplier: Mortech Manufacturing
AH024 AH025 AH026 AH027 AH028 AH029 AH035 AH036 AH037 AH038 AH039 AH040
76549-316EA 423.65 CAD
76549-316 76549-318 76549-320 76549-322 76549-324 76549-326 76549-338 76549-340 76549-342 76549-344 76549-346 76549-348
Disposable Autopsy and Pathology Grossing Prep Knives , Mortech
Knives
Complete disposal of knife and handle eliminates problems of replacement blades fitting and the worries of contamination when working with diseased tissue. Mopec knives have double honed, long-lasting, flexible blades with non-slip handles to ensure accurate dissection.
  • Standard grade Sheffield stainless steel
  • Double honed, flexible blade
  • Pack of ten
  • Set includes two of each knife
All knives are supplied in packs of 10 (an assorted pack containing two of each size is also available). The box in which the knives are packed may be used for archivingMore Product Information