Forceps, Potts-Smith Dressing, Tungsten Carbide, Mortech
Supplier: Mortech Manufacturing
AB401 AB404 AB405 AB406
76548-874EA 598.99 CAD
76548-874 76548-880 76548-882 76548-884
Forceps, Potts-Smith Dressing, Tungsten Carbide, Mortech
Forceps
These forceps are made of extremely high quality tungsten carbide for long lasting use.
  • Tungsten carbide
  • Ribbed grip
  • Serrated tips
Available in a variety of lengths.