Scissors, Tungsten Carbide, Metzenbaum, Mortech
Supplier: Mortech Manufacturing
AA407 AA408 AA409 AA410 AA411 AA412 AA413 AA414 AA415
76548-532EA 567.28 CAD
76548-532 76548-534 76548-536 76548-538 76548-540 76548-542 76548-544 76548-546 76548-548
Scissors, Tungsten Carbide, Metzenbaum, Mortech
Scissors
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Tungsten carbide
  • Narrow, blunt tips
Available with straight or curved edges.