Scissors, Tungsten Carbide, Mortech
Supplier: Mortech Manufacturing
AA401 AA402 AA403 AA404
76548-520EA 567.28 CAD
76548-520 76548-522 76548-524 76548-526
Scissors, Tungsten Carbide, Mortech
Scissors
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Tungsten Carbide
Available with straight or curved edges, and either sharp/sharp or sharp/blunt tips.