Scissors, Mayo Dissection, Curved, Mortech
Supplier: Mortech Manufacturing
AA021 AA022 AA050 AA121 AA122
76548-348EA 65.63 CAD
76548-348 76548-350 76548-392 76548-442 76548-444
Scissors, Mayo Dissection, Curved, Mortech
Scissors
Slightly blunt tips assure precise, controlled and safe post-mortem dissection with minimal damage to surrounding tissue.
  • Stainless steel
  • Curved cutting edge
  • Blunt tips
Available in standard or premium grade.