Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing

Supplier: Saint Gobain Life Sciences
ADK00003 ADK00007 ADK00012 ADK00017 ADK00019 ADK00022 ADK00029 ADK00036 MFLX65102-12 MFLX65102-18
MFLX65102-00EA 241.74 CAD
MFLX65102-00 MFLX65102-02 MFLX65102-04 MFLX65102-06 MFLX65102-08 MFLX65102-10 MFLX65102-14 MFLX65102-16 MFLX65102-12 MFLX65102-18
Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing
Tubing Transfer Tubing
High degree of flexibility, performs well at low temperatures.

  • Global market leader in range of formulations for most extensive chemical compatibility

Formulation: Tygon® E-1000
Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting.
Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent.
Certification: FDA compliant. Made according to GMP.
Temperature range: –67 to 125More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR