Tygon Transfer Tubing, Tygon® E-1000, 1/16" ID x 3/16" OD; 50 Ft
Supplier: Saint Gobain Life Sciences
High degree of flexibility, performs well at low temperatures.
<B>Formulation:</B> Tygon® E-1000<br /><B>Applications:</B> General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting.<br /><B>Characteristics:</B> High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent.<br /><B>Certification:</B> FDA compliant. Made according to GMP.<br /><B>Temperature range:</B> –67 to 125 °F (–55 to 52 °C)<br /><B>Sterilize:</B>Autoclave or ethylene oxide.