CPC® AseptiQuik® G Series Genderless Aseptic Connectors
Supplier: Colder Products
AQG17004 AQG17006 AQG17006HT AQG17104 AQG17106 AQG17108 AQG17112 AQG17MPC AQG17MPCHT(5PK)
MFLX20202-45EA 458.2 CAD
MFLX20202-45 MFLX20202-40 MFLX20202-42 MFLX20202-43 MFLX20202-54 MFLX20202-55 MFLX20202-56 MFLX20202-57 MFLX20202-36 MFLX20202-38
CPC® AseptiQuik® G Series Genderless Aseptic Connectors
Fittings and Connectors
Provide easy, secure sterile connections even in nonsterile environments.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
More Product Information