CPC (Colder) AseptiQuik® G Genderless Connector with MPC Insert, Polycarbonate, High Temperature; 5/PK
Supplier: Colder Products
Provide easy, secure sterile connections even in nonsterile environments.
- Genderless design simplifies system integration—no need to worry about female and male threads
- Robust construction provides reliable performance—even without clamps
- Quick, three-step connection process reduces risk of operator error
- Quick-disconnect fitting features 1/2" ID hose barb connection
- High temperature version allows for gamma and autoclave sterilization
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.<br /><br />Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.<br /><br />All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.