Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
MD-6 MD-6-220V MD-10 MD-10-220V MD-15 MD-15-220V MD-20 MD-20-220V MD-30R(T) MD-30(T)-220V MD-40(T) MD-55(T) MD-55(T) 230V MD-100RLT-115 MFLX72010-55 MFLX72010-70 MFLX72010-75 MFLX72010-85
MFLX72010-00EA 330.22 CAD
MFLX72010-00 MFLX72010-05 MFLX72010-10 MFLX72010-15 MFLX72010-20 MFLX72010-25 MFLX72010-30 MFLX72010-35 MFLX72010-40 MFLX72010-45 MFLX72010-50 MFLX72010-60 MFLX72010-65 MFLX72010-80 MFLX72010-55 MFLX72010-70 MFLX72010-75 MFLX72010-85
Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.