Mag Drive PP Centrifugal Pump w/Enclosed Motor; 5 GPM/11.1 ft, 115V
Supplier: BPH PUMP & EQUIPMENT INC
VWR Item#MFLX72010-20
Your Organization's Price:
Retrieving
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
<b>Delivery information: </b>Includes a 6-ft (1.8 m) cord with stripped ends.