Rapid Response™ COVID-19 Antigen Rapid Test Cassettes - At Home, BTNX
Supplier: BTNX INC CA
COV-19CSHC1 COV-19CSHC2 COV-19CSHC5
CA76540-462CS 2843.92 CAD
CA76540-462 CA76540-464 CA76540-466
Rapid Response™ COVID-19 Antigen Rapid Test Cassettes - At Home, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response® COVID-19 antigen rapid test cassette - at home is an in vitro immunoassay which detects the nucleocapsid proteins of SARS-CoV-2, the virus that causes COVID-19. The test uses nasal swabs taken from asymptomatic or symptomatic people within 7 days of symptom onset.
  • Authorized for self-testing
  • Results in just 10 minutes
  • Compact kit sizes
  • Uses nasal swab samples
  • Easy-to-follow instructions with illustrations
The Rapid Response® COVID-19 antigen rapid test cassette - at home is authorized for home use in a non-laboratory setting with direct anterior nasal (nares) swab samples for: unobserved self-collection for individuals aged 18 years or older, adult-supervised self-collection for individuals age 14 or older, and adults collecting from children two years or older.
This test is authorized under the Health Canada Interim Order (IO #342928).