Masterflex® Direct-Read Variable-Area Flowmeters, Air and Water, Acrylic Housing, Avantor®
Supplier: KEY INSTRUMENTS, LLC
2510A2A00BNBN-CP 2510A2A03BNBN-CP 2510A2A08BNBN-CP 2530A4A67BNBN-CP 2540A5A52PS-CP MFLX32457-00 MFLX32457-02 MFLX32457-06 MFLX32457-14 MFLX32458-07 MFLX32458-08 MFLX32458-10 MFLX32458-16 MFLX32458-18 MFLX32458-22 MFLX32458-26 MFLX32462-00 MFLX32462-02 MFLX32462-04 MFLX32462-10 MFLX32457-12 MFLX32457-22 MFLX32458-00 MFLX32458-02 MFLX32458-06
MFLX32457-10EA 181.7 CAD
MFLX32457-10 MFLX32457-16 MFLX32457-26 MFLX32458-24 MFLX32462-14 MFLX32457-00 MFLX32457-02 MFLX32457-06 MFLX32457-14 MFLX32458-07 MFLX32458-08 MFLX32458-10 MFLX32458-16 MFLX32458-18 MFLX32458-22 MFLX32458-26 MFLX32462-00 MFLX32462-02 MFLX32462-04 MFLX32462-10 MFLX32457-12 MFLX32457-22 MFLX32458-00 MFLX32458-02 MFLX32458-06
Masterflex® Direct-Read Variable-Area Flowmeters, Air and Water, Acrylic Housing, Avantor®
Flow Meters
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct English-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters. Contact our Application Specialists for quotes on acrylic meters with special requirements.