Masterflex® Variable-Area Flowmeter, Direct Read, Acrylic Housing, 4" Scale; 0.4 to 5 GPM Water
Supplier: VWR International
VWR Item#MFLX32458-08
Your Organization's Price:
Retrieving
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct English-Reading Scales
  • Maximum flow rate of 5 GPM
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.<br /><br /><B>Note:</B> There are many additional types of acrylic flowmeters. Contact our Application Specialists for quotes on acrylic meters with special requirements.