Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P04/6-N064-63G P04/6-N064-63ST T54/6-N064-63S P24/6-N064-63G P24/6-N064-63C MFLX03216-88 MFLX03218-39 MFLX03293-38 MFLX03293-40 MFLX03216-40
MFLX03216-36EA 257.54 CAD
MFLX03216-36 MFLX03216-38 MFLX03216-90 MFLX03218-37 MFLX03218-41 MFLX03216-88 MFLX03218-39 MFLX03293-38 MFLX03293-40 MFLX03216-40
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).