Masterflex® Variable-Area Flowmeter with Valve, High-Flow, Brass Housing and Fittings, 65-mm Scale, 58.5 LPM air
Supplier: VWR International
VWR Item#MFLX03293-40
Your Organization's Price:
Retrieving