Fluorescent Labeling Reagents, CypHer5E NHS Ester, Cytiva
Supplier: Cytiva
PA15401
PA15405
CA76492-476EA
823.48
CAD
CA76492-476
CA76492-478
Fluorescent Labeling Reagents, CypHer5E NHS Ester, Cytiva
Electrophoresis Reagents
Western Blotting Reagents
CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH.
- Generic technology can be coupled to any receptor or epitope tag antibody
- Monitors any neutral to acidic event
- Simple fluorescence intensity increase
It is well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.